missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxymethyl-1-methyl-5-nitroimidazole 98.0+%, TCI America™

Catalog No. p-7131045
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H14391G 1 g
H14395G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H14391G Supplier TCI America Supplier No. H14391G
Only null left

Chemical Identifiers

CAS 936-05-0
Molecular Formula C5H7N3O3
Molecular Weight (g/mol) 157.129
MDL Number MFCD00159675
InChI Key JSAQDPJIVQMBAY-UHFFFAOYSA-N
Synonym 1-Methyl-5-nitroimidazole-2-methanol
PubChem CID 557356
IUPAC Name (1-methyl-5-nitroimidazol-2-yl)methanol
SMILES CN1C(=CN=C1CO)[N+](=O)[O-]

Specifications

Melting Point 117°C
Quantity 1 g
Formula Weight 157.13
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Hydroxymethyl-1-methyl-5-nitroimidazole
RTECSNumber NI6801000
TSCA No