missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Hydroxymethyl-1-methyl-5-nitroimidazole 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 117°C |
| Quantity | 1 g |
| Formula Weight | 157.13 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2-Hydroxymethyl-1-methyl-5-nitroimidazole |
Safety and Handling
| RTECSNumber | NI6801000 |
| TSCA | No |