missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxymethyl-1,4-benzodioxane 98.0+%, TCI America™

Catalog No. H080125G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H080125G 25 g
H0801500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H080125G Supplier TCI America Supplier No. H080125G

Chemical Identifiers

CAS 3663-82-9
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00006822
InChI Key GWQOQQVKVOOHTI-UHFFFAOYSA-N
Synonym 2-hydroxymethyl-1,4-benzodioxane, 2,3-dihydro-benzo 1,4 dioxin-2-yl-methanol, 1,4-benzodioxan-2-methanol, 2,3-dihydro-1,4-benzodioxin-2-ylmethanol, 2,3-dihydrobenzo b 1,4 dioxin-2-yl methanol, 1,4-benzodioxane-2-methanol, 2-hydroxymethyl-1,4-benzodioxan, 1,4-benzodioxin-2-methanol, 2,3-dihydro
PubChem CID 19314
IUPAC Name 2,3-dihydro-1,4-benzodioxin-3-ylmethanol
SMILES C1C(OC2=CC=CC=C2O1)CO

Specifications

Melting Point 88°C
Quantity 25 g
Formula Weight 166.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Hydroxymethyl-1,4-benzodioxane
RTECSNumber DF3660000
TSCA No