missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxymethyl-1,4-benzodioxane, 98%

Catalog No. AAA1417006
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1417006 5 g
AAA1417014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1417006 Supplier Thermo Scientific Chemicals Supplier No. A1417006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 3663-82-9 | C9H10O3 | 166.176 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3663-82-9
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00006822
InChI Key GWQOQQVKVOOHTI-UHFFFAOYSA-N
Synonym 2-hydroxymethyl-1,4-benzodioxane, 2,3-dihydro-benzo 1,4 dioxin-2-yl-methanol, 1,4-benzodioxan-2-methanol, 2,3-dihydro-1,4-benzodioxin-2-ylmethanol, 2,3-dihydrobenzo b 1,4 dioxin-2-yl methanol, 1,4-benzodioxane-2-methanol, 2-hydroxymethyl-1,4-benzodioxan, 1,4-benzodioxin-2-methanol, 2,3-dihydro
PubChem CID 19314
IUPAC Name 2,3-dihydro-1,4-benzodioxin-3-ylmethanol
SMILES C1C(OC2=CC=CC=C2O1)CO

Specifications

Melting Point 87°C to 91°C
Quantity 5 g
Beilstein 83736
Formula Weight 166.18
Percent Purity 98%
Chemical Name or Material 2-Hydroxymethyl-1,4-benzodioxane
EINECSNumber 222-921-9
RTECSNumber DF3660000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only