missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxyisophthalaldehyde 98.0+%, TCI America™

Catalog No. H10185G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H10185G 5 g
H10181G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H10185G Supplier TCI America Supplier No. H10185G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3328-69-6
Molecular Formula C8H6O3
Molecular Weight (g/mol) 150.13
MDL Number MFCD00839420
InChI Key JJOPQMAMJLOGFB-UHFFFAOYSA-N
Synonym 2,6-Diformylphenol
PubChem CID 345554
IUPAC Name 2-hydroxybenzene-1,3-dicarbaldehyde
SMILES OC1=C(C=O)C=CC=C1C=O

Specifications

Melting Point 124°C
Quantity 5 g
Formula Weight 150.13
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Hydroxyisophthalaldehyde
TSCA No
Recommended Storage Refrigerator