missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Hydroxyisonicotinic Acid 98.0+%, TCI America™
$124.89 - $499.57
Chemical Identifiers
| CAS | 22282-72-0 |
|---|---|
| Molecular Formula | C6H5NO3 |
| Molecular Weight (g/mol) | 139.11 |
| MDL Number | MFCD01318566 |
| InChI Key | BXHCJLRTXPHUGH-UHFFFAOYSA-N |
| Synonym | 2-hydroxyisonicotinic acid, 2-hydroxy-4-pyridinecarboxylic acid, 2-oxo-1,2-dihydropyridine-4-carboxylic acid, 2-hydroxypyridine-4-carboxylic acid, 2-hydroxy-isonicotinic acid, 2-hydroxyisonicotinicacid, 4-pyridinecarboxylic acid, 1,2-dihydro-2-oxo, pubchem16780, acmc-209yjf, 2-hydroxy isonicotinic acid |
| PubChem CID | 280756 |
| IUPAC Name | 2-oxo-1,2-dihydropyridine-4-carboxylic acid |
| SMILES | OC(=O)C1=CC(=O)NC=C1 |
Chemical Identifiers
| 22282-72-0 | |
| 139.11 | |
| BXHCJLRTXPHUGH-UHFFFAOYSA-N | |
| 280756 | |
| OC(=O)C1=CC(=O)NC=C1 |
| C6H5NO3 | |
| MFCD01318566 | |
| 2-hydroxyisonicotinic acid, 2-hydroxy-4-pyridinecarboxylic acid, 2-oxo-1,2-dihydropyridine-4-carboxylic acid, 2-hydroxypyridine-4-carboxylic acid, 2-hydroxy-isonicotinic acid, 2-hydroxyisonicotinicacid, 4-pyridinecarboxylic acid, 1,2-dihydro-2-oxo, pubchem16780, acmc-209yjf, 2-hydroxy isonicotinic acid | |
| 2-oxo-1,2-dihydropyridine-4-carboxylic acid |
Specifications
| 22282-72-0 | |
| White-Yellow | |
| MFCD01318566 | |
| 2-hydroxyisonicotinic acid, 2-hydroxy-4-pyridinecarboxylic acid, 2-oxo-1,2-dihydropyridine-4-carboxylic acid, 2-hydroxypyridine-4-carboxylic acid, 2-hydroxy-isonicotinic acid, 2-hydroxyisonicotinicacid, 4-pyridinecarboxylic acid, 1,2-dihydro-2-oxo, pubchem16780, acmc-209yjf, 2-hydroxy isonicotinic acid | |
| OC(=O)C1=CC(=O)NC=C1 | |
| 139.11 | |
| 139.11 | |
| Crystalline Powder |
| 328°C | |
| C6H5NO3 | |
| 5 g | |
| BXHCJLRTXPHUGH-UHFFFAOYSA-N | |
| 2-oxo-1,2-dihydropyridine-4-carboxylic acid | |
| 280756 | |
| ≥98.0% (GC) | |
| 2-Hydroxyisonicotinic Acid |
Safety and Handling
TSCA : No