missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Hydroxyisonicotinic Acid 98.0+%, TCI America™
Spécifications
Spécifications
| Melting Point | 328°C |
| Color | White-Yellow |
| Quantity | 5 g |
| Formula Weight | 139.11 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2-Hydroxyisonicotinic Acid |
Sécurité et manipulation
| TSCA | No |