Learn More
2-Hydroxyisobutyric acid, 99% (dry wt.), water <2%
Description
2-Hydroxyisobutyric acid is used as an electrolyte in the separation of the metal ions by on-line cyclic voltammetry. It is also used as a building block for the synthesis of polymers. Its conjugate base, 2-hydroxybutyrate is utilized to catabolize L-threonine as well as synthesize glutathione. Further, it is used as an indicator for the early detection insulin resistance in non-diabetic subjects.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2-Hydroxyisobutyric acid is used as an electrolyte in the separation of the metal ions by on-line cyclic voltammetry. It is also used as a building block for the synthesis of polymers. Its conjugate base, 2-hydroxybutyrate is utilized to catabolize L-threonine as well as synthesize glutathione. Further, it is used as an indicator for the early detection insulin resistance in non-diabetic subjects.
Solubility
Soluble in water, ether, alcohol, methanol and hot benzene.
Notes
Incompatible with oxidizing agents, bases and reducing agents.
Specifications
Specifications
| Melting Point | 77°C to 80°C |
| Density | 1.24 |
| Boiling Point | 114°C (12 mmHg) |
| Quantity | 25 g |
| Beilstein | 1744739 |
| Sensitivity | Hygroscopic |
| Solubility Information | Soluble in water,ether,alcohol,methanol and hot benzene. |
| Formula Weight | 104.11 |
| Percent Purity | 99% |
| Assay | (dry wt.), water <2% |
| Show More |
Safety and Handling
| Hazard Category | H302-H315-H318-H335 |
| Hazard Statement | GHS H Statement H318-H335-H315 Causes serious eye damage. May cause respiratory irritation. Causes skin irritation. |
| Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c |
| EINECSNumber | 209-848-8 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only