missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Hydroxyethyl stearate
CAS: 111-60-4 | C20H40O3 | 328.53 g/mol
$59.48 - $112.00
Chemical Identifiers
| CAS | 111-60-4 |
|---|---|
| Molecular Formula | C20H40O3 |
| Molecular Weight (g/mol) | 328.53 |
| MDL Number | MFCD00051465 |
| InChI Key | RFVNOJDQRGSOEL-UHFFFAOYSA-N |
| Synonym | glycol stearate, cremophor a, 2-hydroxyethyl stearate, polyethylene glycol monostearate, ethylene glycol monostearate, cerasynt m, clearate g, cerasynt mn, cithrol ps, polyoxyl 40 stearate |
| PubChem CID | 24762 |
| IUPAC Name | 2-hydroxyethyl octadecanoate |
| SMILES | CCCCCCCCCCCCCCCCCC(=O)OCCO |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC346951000
|
Thermo Scientific Chemicals
346951000 |
100 g | Glass bottle |
Each for $59.48
|
|
||||
|
AC346952500
|
Thermo Scientific Chemicals
346952500 |
250 g | Glass bottle |
Each for $112.00
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 111-60-4 | |
| 328.53 | |
| RFVNOJDQRGSOEL-UHFFFAOYSA-N | |
| 24762 | |
| CCCCCCCCCCCCCCCCCC(=O)OCCO |
| C20H40O3 | |
| MFCD00051465 | |
| glycol stearate, cremophor a, 2-hydroxyethyl stearate, polyethylene glycol monostearate, ethylene glycol monostearate, cerasynt m, clearate g, cerasynt mn, cithrol ps, polyoxyl 40 stearate | |
| 2-hydroxyethyl octadecanoate |
Specifications
| 111-60-4 | |
| White to Yellow | |
| Authentic | |
| C20H40O3 | |
| 100 g | |
| Solubility in water: insoluble in h2o | |
| CCCCCCCCCCCCCCCCCC(=O)OCCO | |
| 328.53 | |
| 328.53 | |
| compound is a mixture of various chain-length esters and di(ethylene glycol) ester | |
| 2-Hydroxyethyl stearate |
| 55°C to 60°C | |
| 149°C | |
| Glass bottle | |
| MFCD00051465 | |
| glycol stearate, cremophor a, 2-hydroxyethyl stearate, polyethylene glycol monostearate, ethylene glycol monostearate, cerasynt m, clearate g, cerasynt mn, cithrol ps, polyoxyl 40 stearate | |
| RFVNOJDQRGSOEL-UHFFFAOYSA-N | |
| 2-hydroxyethyl octadecanoate | |
| 24762 | |
| Technical | |
| Flakes |
Safety and Handling
EINECSNumber : 203-886-9
RTECSNumber : WI4100000
TSCA : TSCA
RUO – Research Use Only