missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxyethyl Starch, Spectrum™ Chemical

Catalog No. p-7157574
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Poly Bottle
Poly Pail
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18600112 250 g Poly Bottle
18600113 1 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18600112 Supplier Spectrum Chemical Mfg Cor Supplier No. H3012250GM
Only null left

H3012, 9005-27-0

Spectrum™ Chemical 2-Hydroxyethyl Starch

Chemical Identifiers

CAS 9005-27-0

Specifications

CAS Max % 100%
Packaging Poly Bottle
Quantity 250 g
Grade Reagent