missing translation for 'onlineSavingsMsg'
Learn More

(2-Hydroxyethyl)dimethyl(3-sulfopropyl)ammonium Hydroxide Inner Salt 98.0+%, TCI America™

Catalog No. H13991G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H13991G 1 g
H13995G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H13991G Supplier TCI America Supplier No. H13991G

Chemical Identifiers

CAS 38880-58-9
Molecular Formula C7H18NO4S+
Molecular Weight (g/mol) 212.284
MDL Number MFCD01320397
InChI Key CNXPCGBLGHKAIL-UHFFFAOYSA-O
Synonym 3-[(2-Hydroxyethyl)dimethylammonio]propane-1-sulfonate, NDSB 211
PubChem CID 102269415
IUPAC Name 3-[dimethyl(2-oxonioethyl)azaniumyl]propane-1-sulfonate
SMILES C[N+](C)(CCCS(=O)(=O)[O-])CC[OH2+]

Specifications

Melting Point 248°C
Color White
Quantity 1 g
Formula Weight 211.28
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material (2-Hydroxyethyl)dimethyl(3-sulfopropyl)ammonium Hydroxide Inner Salt [for Biochemical Research]