missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxyethyl Acrylate (stabilized with MEHQ) 95.0+%, TCI America™

Catalog No. A074325ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A074325ML 25 mL
A0743500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A074325ML Supplier TCI America Supplier No. A074325ML

Chemical Identifiers

CAS 818-61-1
Molecular Formula C5H8O3
Molecular Weight (g/mol) 116.116
MDL Number MFCD00002865
InChI Key OMIGHNLMNHATMP-UHFFFAOYSA-N
Synonym 2-hydroxyethyl acrylate, hydroxyethyl acrylate, 2-propenoic acid, 2-hydroxyethyl ester, ethylene glycol monoacrylate, bisomer 2hea, acrylic acid 2-hydroxyethyl ester, 2-acryloyloxy ethanol, ethylene glycol, acrylate, acrylic acid, 2-hydroxyethyl ester, 2-hydroxyethylacrylate
PubChem CID 13165
IUPAC Name 2-hydroxyethyl prop-2-enoate
SMILES C=CC(=O)OCCO

Specifications

Color Yellow
Boiling Point 92°C
Quantity 25 mL
UN Number 2922
Formula Weight 116.12
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Hydroxyethyl Acrylate (stabilized with MEHQ)
EINECSNumber (2)-0995
RTECSNumber AT1750000
TSCA Yes