missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxycyclohexanone dimer, 90+%

Catalog No. AAH2719903
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2719903 1 g
AAH2719906 5 g
AAH2719914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH2719903 Supplier Thermo Scientific Chemicals Supplier No. H2719903
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 30282-14-5 | C6H10O2 | 114.144 g/mol

It is a reactant involved in the synthesis of substrates for tin tetrachloride mediated fragmentation to produce ynones and ynoates, tandem oxidation coupling reactions with diamines, asymmetric allylic alkylation of enolates and formate reduction of allylic carbonates for polypropionate systems.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a reactant involved in the synthesis of substrates for tin tetrachloride mediated fragmentation to produce ynones and ynoates, tandem oxidation coupling reactions with diamines, asymmetric allylic alkylation of enolates and formate reduction of allylic carbonates for polypropionate systems.

Solubility
Partly miscible in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep away from ignition sources. Protect from electrostatic charges. Ensure adequate ventilation.

Chemical Identifiers

CAS 30282-14-5
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.144
MDL Number MFCD00151157
InChI Key ODZTXUXIYGJLMC-UHFFFAOYSA-N
Synonym 2-hydroxycyclohexanone, adipoin, 2-hydroxy-1-cyclohexanone, cyclohexanone, 2-hydroxy, 2-hydroxy-cyclohexanone, cyclohexanone, 2-hydroxy-, 2r, cyclohexanone, 2-hydroxy-, dimer, cyclohexanone, dimer, zlchem 1128, acmc-20n5uh
PubChem CID 10785
ChEBI CHEBI:17878
IUPAC Name 2-hydroxycyclohexan-1-one
SMILES C1CCC(=O)C(C1)O

Specifications

Melting Point 100°C to 107°C
Boiling Point 83°C to 86°C (13 mmHg)
Flash Point 79°C (174°F)
Quantity 1 g
UN Number UN1325
Solubility Information Partly miscible in water.
Formula Weight 228.28
Percent Purity ≥90%
Chemical Name or Material 2-Hydroxycyclohexanone dimer
Hazard Category H228-H315-H319-H335
Hazard Statement GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only