missing translation for 'onlineSavingsMsg'
Learn More

2'-Hydroxychalcone 98.0+%, TCI America™

Supplier:  TCI America H038525G

Catalog No. H038525G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2′-Hydroxychalcone
88°C
C15H12O2
25 g
AETKQQBRKSELEL-ZHACJKMWSA-N
(E)-1-(2-hydroxyphenyl)-3-phenylprop-2-en-1-one
638276
224.26
Crystalline Powder
1214-47-7
Yellow
MFCD00016441
2'-hydroxychalcone, o-hydroxychalcone, e-2'-hydroxychalcone, 1-2-hydroxyphenyl-3-phenylprop-2-en-1-one, chalcone, 2'-hydroxy, ccris 7796, 1-2-hydroxyphenyl-3-phenyl-2-propen-1-one, unii-vy06dz94oc, acrylophenone, 2'-hydroxy-3-phenyl, 2-propen-1-one, 1-2-hydroxyphenyl-3-phenyl
C1=CC=C(C=C1)C=CC(=O)C2=CC=CC=C2O
224.259
CHEBI:27916
≥98.0% (HPLC)

Chemical Identifiers

1214-47-7
224.259
AETKQQBRKSELEL-ZHACJKMWSA-N
638276
(E)-1-(2-hydroxyphenyl)-3-phenylprop-2-en-1-one
C15H12O2
MFCD00016441
2'-hydroxychalcone, o-hydroxychalcone, e-2'-hydroxychalcone, 1-2-hydroxyphenyl-3-phenylprop-2-en-1-one, chalcone, 2'-hydroxy, ccris 7796, 1-2-hydroxyphenyl-3-phenyl-2-propen-1-one, unii-vy06dz94oc, acrylophenone, 2'-hydroxy-3-phenyl, 2-propen-1-one, 1-2-hydroxyphenyl-3-phenyl
CHEBI:27916
C1=CC=C(C=C1)C=CC(=O)C2=CC=CC=C2O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : UD5579400

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions