missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxyacetophenone 98.0+%, TCI America™

Catalog No. H03475G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H03475G 5 g
H034725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H03475G Supplier TCI America Supplier No. H03475G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 582-24-1
Molecular Formula C8H8O2
Molecular Weight (g/mol) 136.15
MDL Number MFCD00041829
InChI Key ZWVHTXAYIKBMEE-UHFFFAOYSA-N
Synonym 2-hydroxyacetophenone, benzoylcarbinol, glycolophenone, phenacyl alcohol, alpha-hydroxyacetophenone, 2-hydroxy-1-phenylethan-1-one, ethanone, 2-hydroxy-1-phenyl, methanol, benzoyl, omega-hydroxyacetophenone, acetophenone, 2-hydroxy
PubChem CID 68490
ChEBI CHEBI:28341
IUPAC Name 2-hydroxy-1-phenylethanone
SMILES C1=CC=C(C=C1)C(=O)CO

Specifications

Melting Point 88°C
Color White-Yellow
Boiling Point 126°C
Quantity 5 g
Formula Weight 136.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Hydroxyacetophenone
EINECSNumber (3)-4208
TSCA Yes