missing translation for 'onlineSavingsMsg'
Learn More

2'-Hydroxyacetophenone 98.0+%, TCI America™

Catalog No. H0192500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H0192500G 500 g
H019225G 25 g
H0192100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H0192500G Supplier TCI America Supplier No. H0192500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 118-93-4
Molecular Formula C8H8O2
Molecular Weight (g/mol) 136.15
MDL Number MFCD00002219
InChI Key JECYUBVRTQDVAT-UHFFFAOYSA-N
Synonym 2'-hydroxyacetophenone, 2-acetylphenol, 1-2-hydroxyphenyl ethanone, o-hydroxyacetophenone, o-acetylphenol, o-hydroxyphenyl methyl ketone, ethanone, 1-2-hydroxyphenyl, 1-2-hydroxyphenyl ethan-1-one, acetophenone, 2'-hydroxy, 2-hydroxyphenyl methyl ketone
PubChem CID 8375
IUPAC Name 1-(2-hydroxyphenyl)ethanone
SMILES CC(=O)C1=CC=CC=C1O

Specifications

Melting Point 6°C
Color Yellow
Boiling Point 218°C
Quantity 500 g
Formula Weight 136.15
Percent Purity ≥98.0% (HPLC,T)
Physical Form Liquid
Chemical Name or Material 2′-Hydroxyacetophenone
RTECSNumber AM8575000
TSCA Yes