missing translation for 'onlineSavingsMsg'
Learn More

2'-Hydroxyacetophenone, 98%

Catalog No. AAA1255036
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1255036 500 g
AAA1255022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1255036 Supplier Thermo Scientific Chemicals Supplier No. A1255036
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 118-93-4 | C8H8O2 | 136.15 g/mol

2'-Hydroxyacetophenone is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′-Hydroxyacetophenone is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 118-93-4
Molecular Formula C8H8O2
Molecular Weight (g/mol) 136.15
MDL Number MFCD00002219
InChI Key JECYUBVRTQDVAT-UHFFFAOYSA-N
Synonym 2'-hydroxyacetophenone, 2-acetylphenol, 1-2-hydroxyphenyl ethanone, o-hydroxyacetophenone, o-acetylphenol, o-hydroxyphenyl methyl ketone, ethanone, 1-2-hydroxyphenyl, 1-2-hydroxyphenyl ethan-1-one, acetophenone, 2'-hydroxy, 2-hydroxyphenyl methyl ketone
PubChem CID 8375
IUPAC Name 1-(2-hydroxyphenyl)ethanone
SMILES CC(=O)C1=CC=CC=C1O

Specifications

Melting Point 3°C to 6°C
Density 1.13
Boiling Point 218°C
Flash Point 106°C (222°F)
Refractive Index 1.5584
Quantity 500 g
Beilstein 386123
Solubility Information Slightly soluble in water.
Formula Weight 136.15
Percent Purity 98%
Chemical Name or Material 2'-Hydroxyacetophenone
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 204-288-0
RTECSNumber AM8575000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only