missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-6-methylpyridine, 98%

Catalog No. AAA1476414
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1476414 25 g
AAA1476406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1476414 Supplier Thermo Scientific Chemicals Supplier No. A1476414
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 3279-76-3 | C6H7NO | 109.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3279-76-3
Molecular Formula C6H7NO
Molecular Weight (g/mol) 109.13
MDL Number MFCD00006279
InChI Key JEAVIRYCMBDJIU-UHFFFAOYSA-N
Synonym 2-hydroxy-6-methylpyridine, 6-methylpyridin-2-ol, 6-methylpyridin-2 1h-one, 6-methyl-2-pyridinol, 6-methyl-2-pyridone, 6-methyl-2-pyridinone, 2 1h-pyridinone, 6-methyl, 6-methyl-2 1h-pyridinone, 2-hydroxy-6-picoline, 6-hydroxy-2-picoline
PubChem CID 76772
IUPAC Name 6-methyl-1H-pyridin-2-one
SMILES CC1=CC=CC(=O)N1

Specifications

Melting Point 157°C to 161°C
Odor Odorless
Quantity 25 g
Beilstein 107073
Formula Weight 109.13
Percent Purity 98%
Chemical Name or Material 2-Hydroxy-6-methylpyridine
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
EINECSNumber 221-919-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only