missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-6-methylpyridine, 97%

Catalog No. p-145976
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC156390050 5 g Glass bottle
AC156390250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC156390050 Supplier Thermo Scientific Chemicals Supplier No. 156390050
Only null left

CAS: 3279-76-3 | C6H7NO | 109.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3279-76-3
Molecular Formula C6H7NO
Molecular Weight (g/mol) 109.13
MDL Number MFCD00006279
InChI Key JEAVIRYCMBDJIU-UHFFFAOYSA-N
Synonym 2-hydroxy-6-methylpyridine, 6-methylpyridin-2-ol, 6-methylpyridin-2 1h-one, 6-methyl-2-pyridinol, 6-methyl-2-pyridone, 6-methyl-2-pyridinone, 2 1h-pyridinone, 6-methyl, 6-methyl-2 1h-pyridinone, 2-hydroxy-6-picoline, 6-hydroxy-2-picoline
PubChem CID 76772
IUPAC Name 6-methyl-1H-pyridin-2-one
SMILES CC1=CC=CC(=O)N1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 155°C to 160°C
Color White to Cream
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula (C5H4N)CH2OH
Quantity 5 g
Beilstein 21, 49
Formula Weight 109.13
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2-Hydroxy-6-methylpyridine, 97%
Show More Show Less
EINECSNumber 221-919-5
TSCA TSCA

RUO – Research Use Only