missing translation for 'onlineSavingsMsg'
Learn More

2'-Hydroxy-6'-methoxyacetophenone, 97%

Catalog No. aaa1853914
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1853914 25 g
AAA1853903 1 g
AAA1853906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1853914 Supplier Thermo Scientific Chemicals Supplier No. A1853914
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 703-23-1 | C9H10O3 | 166.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 703-23-1
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.18
MDL Number MFCD00008732
InChI Key UENLHUMCIOWYQN-UHFFFAOYSA-N
PubChem CID 69709
IUPAC Name 1-(2-hydroxy-6-methoxyphenyl)ethanone
SMILES COC1=CC=CC(O)=C1C(C)=O

Specifications

Melting Point 56°C to 59°C
Boiling Point 141°C (16 mmHg)
Quantity 25 g
Beilstein 1869049
Synonym 1-2-hydroxy-6-methoxyphenyl ethanone, 2'-hydroxy-6'-methoxyacetophenone, 2-hydroxy-6-methoxyacetophenone, 1-2-hydroxy-6-methoxyphenyl ethan-1-one, ethanone, 1-2-hydroxy-6-methoxyphenyl, acetophenone, 2'-hydroxy-6'-methoxy, unii-m5vqg8968n, ethanone,1-2-hydroxy-6-methoxyphenyl, 1-2-hydroxy-6-methoxy-phenyl-ethanone, 1-acetyl-2-hydroxy-6-methoxybenzene
Formula Weight 166.18
Percent Purity 97%
Chemical Name or Material 2'-Hydroxy-6'-methoxyacetophenone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 211-872-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only