missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-5-(trifluoromethoxy)benzaldehyde, 99%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 398200010

 View more versions of this product

Catalog No. AC398200010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

2-Hydroxy-5-(trifluoromethoxy)benzaldehyde
98.5
82.0°C (60.0 mmHg)
98.5% min. (HPLC)
C8H5F3O3
MFCD00075249
2-hydroxy-5-trifluoromethoxy benzaldehyde, 5-trifluoromethoxy salicylaldehyde, benzaldehyde, 2-hydroxy-5-trifluoromethoxy, acmc-20antc, 5-trifluoromethoxysalicylaldehyde, 5-trifluoromethoxy-salicylaldehyde, 2-hydroxy-5-trifluoromethoxy-benzaldehyde, 20hydroxy-5-trifluoromethoxy benzaldehyde, 2-oxidanyl-5-trifluoromethyloxy benzaldehyde, benzaldehyde,2-hydroxy-5-trifluoromethoxy
C1=CC(=C(C=C1OC(F)(F)F)C=O)O
206.12
206.12
93249-62-8
100.0
Authentic
Glass bottle
CF3OC6H3(OH)CHO
1g
WQUZBERVMUEJTD-UHFFFAOYSA-N
2-hydroxy-5-(trifluoromethoxy)benzaldehyde
1268058
99%

Chemical Identifiers

93249-62-8
206.12
WQUZBERVMUEJTD-UHFFFAOYSA-N
1268058
C1=CC(=C(C=C1OC(F)(F)F)C=O)O
C8H5F3O3
MFCD00075249
2-hydroxy-5-trifluoromethoxy benzaldehyde, 5-trifluoromethoxy salicylaldehyde, benzaldehyde, 2-hydroxy-5-trifluoromethoxy, acmc-20antc, 5-trifluoromethoxysalicylaldehyde, 5-trifluoromethoxy-salicylaldehyde, 2-hydroxy-5-trifluoromethoxy-benzaldehyde, 20hydroxy-5-trifluoromethoxy benzaldehyde, 2-oxidanyl-5-trifluoromethyloxy benzaldehyde, benzaldehyde,2-hydroxy-5-trifluoromethoxy
2-hydroxy-5-(trifluoromethoxy)benzaldehyde
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only