missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-5-nitropyridine 97.0+%, TCI America™

Catalog No. H067325G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H067325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H067325G Supplier TCI America Supplier No. H067325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

High-purity heterocyclic building block with >97.0% purity for chemical research applications.

Specifications

CAS 5418-51-9
Melting Point 186°C
Molecular Formula C5H4N2O3
Quantity 25 g
MDL Number MFCD00006276
Synonym 2-hydroxy-5-nitropyridine, 5-nitropyridin-2-ol, 5-nitro-2-pyridinol, 2 1h-pyridinone, 5-nitro, 5-nitropyridin-2 1h-one, 5-nitro-2-hydroxypyridine, 5-nitro-2-pyridone, 5-nitro-1,2-dihydropyridin-2-one, 5-nitro-2-pyridol, pubchem1205
InChI Key XKWSQIMYNVLGBO-UHFFFAOYSA-N
SMILES C1=CC(=O)NC=C1[N+](=O)[O-]
IUPAC Name 5-nitro-1H-pyridin-2-one
Molecular Weight (g/mol) 140.098
PubChem CID 79453
Formula Weight 140.10
Percent Purity ≥97.0% (GC,T)
Grade Not Available
Physical Form Crystalline Powder
Chemical Name or Material 2-Hydroxy-5-nitropyridine
Show More Show Less