missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-5-nitropyridine, 98+%

CAS: 5418-51-9 | C5H4N2O3 | 140.098 g/mol

Supplier:  Thermo Scientific Chemicals A1392114

 View more versions of this product

Catalog No. AAA1392114


Only null left
Explore available promotions

Description

Description

A comparison of catalysts promotes imidazolide couplings including the identification of 2-hydroxy-5-nitropyridine as a new, safe, and effective catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A comparison of catalysts promotes imidazolide couplings including the identification of 2-hydroxy-5-nitropyridine as a new, safe, and effective catalyst.

Solubility
Soluble in hot water and alkali liquor, Insoluble in most of organic solvents.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Specifications

2-Hydroxy-5-nitropyridine
188°C to 191°C
MFCD00006276
120352
Soluble in hot water and alkali liquor,Insoluble in most of organic solvents.
C1=CC(=O)NC=C1[N+](=O)[O-]
140.098
140.1
5418-51-9
C5H4N2O3
25 g
2-hydroxy-5-nitropyridine, 5-nitropyridin-2-ol, 5-nitro-2-pyridinol, 2 1h-pyridinone, 5-nitro, 5-nitropyridin-2 1h-one, 5-nitro-2-hydroxypyridine, 5-nitro-2-pyridone, 5-nitro-1,2-dihydropyridin-2-one, 5-nitro-2-pyridol, pubchem1205
XKWSQIMYNVLGBO-UHFFFAOYSA-N
5-nitro-1H-pyridin-2-one
79453
≥98%

Chemical Identifiers

5418-51-9
140.098
XKWSQIMYNVLGBO-UHFFFAOYSA-N
79453
C1=CC(=O)NC=C1[N+](=O)[O-]
C5H4N2O3
MFCD00006276
2-hydroxy-5-nitropyridine, 5-nitropyridin-2-ol, 5-nitro-2-pyridinol, 2 1h-pyridinone, 5-nitro, 5-nitropyridin-2 1h-one, 5-nitro-2-hydroxypyridine, 5-nitro-2-pyridone, 5-nitro-1,2-dihydropyridin-2-one, 5-nitro-2-pyridol, pubchem1205
5-nitro-1H-pyridin-2-one
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 226-525-7

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only