missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Hydroxy-5-methylpyridine 98.0+%, TCI America™
Supplier: TCI America H116725G
Specifications
| 2-Hydroxy-5-methylpyridine | |
| 184°C | |
| 25 g | |
| SOHMZGMHXUQHGE-UHFFFAOYSA-N | |
| 5-methyl-1H-pyridin-2-one | |
| 70482 | |
| ≥98.0% (GC,T) |
| 1003-68-5 | |
| C6H7NO | |
| 2-hydroxy-5-methylpyridine, 5-methylpyridin-2-ol, 2-hydroxy-5-picoline, 5-methyl-2-pyridinol, 5-methylpyridin-2 1h-one, 5-methyl-2-pyridone, 5-methyl-2 1h-pyridinone, 2 1h-pyridinone, 5-methyl, 2-pyridinol, 5-methyl, 6-hydroxy-3-picoline | |
| CC1=CNC(=O)C=C1 | |
| 109.128 | |
| 109.13 | |
| Crystalline Powder |
Chemical Identifiers
| 1003-68-5 | |
| 109.128 | |
| 2-hydroxy-5-methylpyridine, 5-methylpyridin-2-ol, 2-hydroxy-5-picoline, 5-methyl-2-pyridinol, 5-methylpyridin-2 1h-one, 5-methyl-2-pyridone, 5-methyl-2 1h-pyridinone, 2 1h-pyridinone, 5-methyl, 2-pyridinol, 5-methyl, 6-hydroxy-3-picoline | |
| 5-methyl-1H-pyridin-2-one |
| C6H7NO | |
| SOHMZGMHXUQHGE-UHFFFAOYSA-N | |
| 70482 | |
| CC1=CNC(=O)C=C1 |
Safety and Handling
TSCA : No