missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-5-methylpyridine 98.0+%, TCI America™

Catalog No. H11675G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H11675G 5 g
H116725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H11675G Supplier TCI America Supplier No. H11675G

Chemical Identifiers

CAS 1003-68-5
Molecular Formula C6H7NO
Molecular Weight (g/mol) 109.128
InChI Key SOHMZGMHXUQHGE-UHFFFAOYSA-N
Synonym 2-hydroxy-5-methylpyridine, 5-methylpyridin-2-ol, 2-hydroxy-5-picoline, 5-methyl-2-pyridinol, 5-methylpyridin-2 1h-one, 5-methyl-2-pyridone, 5-methyl-2 1h-pyridinone, 2 1h-pyridinone, 5-methyl, 2-pyridinol, 5-methyl, 6-hydroxy-3-picoline
PubChem CID 70482
IUPAC Name 5-methyl-1H-pyridin-2-one
SMILES CC1=CNC(=O)C=C1

Specifications

Melting Point 184°C
Quantity 5 g
Formula Weight 109.13
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Hydroxy-5-methylpyridine