missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-5-methylpyridine, 98%

Catalog No. AAL1952814
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1952814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1952814 Supplier Thermo Scientific Chemicals Supplier No. L1952814
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1003-68-5 | C6H7NO | 109.128 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1003-68-5
Molecular Formula C6H7NO
Molecular Weight (g/mol) 109.128
MDL Number MFCD03092888
InChI Key SOHMZGMHXUQHGE-UHFFFAOYSA-N
Synonym 2-hydroxy-5-methylpyridine, 5-methylpyridin-2-ol, 2-hydroxy-5-picoline, 5-methyl-2-pyridinol, 5-methylpyridin-2 1h-one, 5-methyl-2-pyridone, 5-methyl-2 1h-pyridinone, 2 1h-pyridinone, 5-methyl, 2-pyridinol, 5-methyl, 6-hydroxy-3-picoline
PubChem CID 70482
IUPAC Name 5-methyl-1H-pyridin-2-one
SMILES CC1=CNC(=O)C=C1

Specifications

Melting Point 183°C to 187°C
Odor Odorless
Quantity 25 g
Beilstein 107074
Formula Weight 109.13
Percent Purity 98%
Chemical Name or Material 2-Hydroxy-5-methylpyridine
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
EINECSNumber 213-719-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only