missing translation for 'onlineSavingsMsg'
Learn More

2'-Hydroxy-5'-methylacetophenone 98.0+%, TCI America™

Numéro de catalogue. p-7130724
Change view
Click to view available options
Quantity:
5 g
25 g
Numéro de catalogue. Quantity
H07905G 5 g
H079025G 25 g
2 options

Numéro de catalogue. H07905G

Supplier: TCI America H07905G

Il en reste null

Identifiants chimiques

CAS 1450-72-2
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.177
MDL Number MFCD00002380
InChI Key YNPDFBFVMJNGKZ-UHFFFAOYSA-N
Synonym 2'-hydroxy-5'-methylacetophenone, 1-2-hydroxy-5-methylphenyl ethanone, 2-hydroxy-5-methylacetophenone, o-acetyl-p-cresol, ethanone, 1-2-hydroxy-5-methylphenyl, 2-acetyl-4-methylphenol, 1-2-hydroxy-5-methylphenyl ethan-1-one, 1-hydroxy-2-acetyl-4-methylbenzene, 1-2-hydroxy-5-methyl-phenyl-ethanone, unii-11661u1zen
PubChem CID 15068
IUPAC Name 1-(2-hydroxy-5-methylphenyl)ethanone
SMILES CC1=CC(=C(C=C1)O)C(=O)C

Spécifications

Melting Point 47°C
Color Yellow
Quantity 5 g
Formula Weight 150.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2′-Hydroxy-5′-methylacetophenone
EINECSNumber (3)-2937
TSCA Yes