missing translation for 'onlineSavingsMsg'
Learn More

2'-Hydroxy-5'-methylacetophenone, 98%

Catalog No. AAA1313614
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1313614 25 g
AAA1313606 5 g
AAA1313622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1313614 Supplier Thermo Scientific Chemicals Supplier No. A1313614
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1450-72-2 | C9H10O2 | 150.177 g/mol

2'-Hydroxy-5'-methylacetophenone is used to produce 2-ethyl-4-methyl-phenol. It is used as an intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′-Hydroxy-5′-methylacetophenone is used to produce 2-ethyl-4-methyl-phenol. It is used as an intermediate in organic synthesis.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 1450-72-2
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.177
MDL Number MFCD00002380
InChI Key YNPDFBFVMJNGKZ-UHFFFAOYSA-N
Synonym 2'-hydroxy-5'-methylacetophenone, 1-2-hydroxy-5-methylphenyl ethanone, 2-hydroxy-5-methylacetophenone, o-acetyl-p-cresol, ethanone, 1-2-hydroxy-5-methylphenyl, 2-acetyl-4-methylphenol, 1-2-hydroxy-5-methylphenyl ethan-1-one, 1-hydroxy-2-acetyl-4-methylbenzene, 1-2-hydroxy-5-methyl-phenyl-ethanone, unii-11661u1zen
PubChem CID 15068
IUPAC Name 1-(2-hydroxy-5-methylphenyl)ethanone
SMILES CC1=CC(=C(C=C1)O)C(=O)C

Specifications

Melting Point 45°C to 48°C
Density 1.079
Boiling Point 112°C to 114°C (12 mmHg)
Flash Point >110°C (230°F)
Quantity 25 g
Beilstein 1364840
Solubility Information Insoluble in water.
Formula Weight 150.18
Percent Purity 98%
Chemical Name or Material 2'-Hydroxy-5'-methylacetophenone
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 215-915-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only