missing translation for 'onlineSavingsMsg'
Learn More

2'-Hydroxy-5'-methyl-3'-nitroacetophenone 98.0+%, TCI America™

Catalog No. H14795G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H14795G 5 g
H147925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H14795G Supplier TCI America Supplier No. H14795G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 66108-30-3
Molecular Formula C9H9NO4
Molecular Weight (g/mol) 195.17
MDL Number MFCD00192216
InChI Key XSHQMMIEZHWNAK-UHFFFAOYSA-N
Synonym 2'-hydroxy-5'-methyl-3'-nitroacetophenone, 1-2-hydroxy-5-methyl-3-nitrophenyl ethanone, 2-hydroxy-5-methyl-3-nitroacetophenone, 2-acetyl-4-methyl-6-nitrophenol, ethanone,1-2-hydroxy-5-methyl-3-nitrophenyl, 1-acetyl-2-hydroxy-5-methyl-3-nitrobenzene, 2'-hydroxy-3'-nitro-5'-methylacetophenone, 1-2-hydroxy-5-methyl-3-nitro-phenyl-ethanone, 1-5-methyl-3-nitro-2-oxidanyl-phenyl ethanone, 1-2-hydroxy-5-methyl-3-nitrophenyl-1-ethanone
PubChem CID 736162
IUPAC Name 1-(2-hydroxy-5-methyl-3-nitrophenyl)ethan-1-one
SMILES CC(=O)C1=CC(C)=CC(=C1O)[N+]([O-])=O

Specifications

Melting Point 134°C
Color Yellow
Quantity 5 g
Formula Weight 195.17
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2′-Hydroxy-5′-methyl-3′-nitroacetophenone