missing translation for 'onlineSavingsMsg'
Learn More

2'-Hydroxy-5'-methoxyacetophenone, 97%

Catalog No. AAA1921606
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1921606 5 g
AAA1921614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1921606 Supplier Thermo Scientific Chemicals Supplier No. A1921606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 705-15-7 | C9H10O3 | 166.176 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 705-15-7
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00008731
InChI Key MLIBGOFSXXWRIY-UHFFFAOYSA-N
Synonym 2'-hydroxy-5'-methoxyacetophenone, 1-2-hydroxy-5-methoxyphenyl ethanone, 2-hydroxy-5-methoxyacetophenone, 1-2-hydroxy-5-methoxyphenyl ethan-1-one, ethanone, 1-2-hydroxy-5-methoxyphenyl, 5-methoxy-2-hydroxyacetophenone, acetophenone, 2'-hydroxy-5'-methoxy, 2'-hydroxy-5'-methoxyacetylphenone, 1-2-hydroxy-5-methoxy-phenyl-ethanone, 1-acetyl-2-hydroxy-5-methoxybenzene
PubChem CID 69714
IUPAC Name 1-(2-hydroxy-5-methoxyphenyl)ethanone
SMILES CC(=O)C1=C(C=CC(=C1)OC)O

Specifications

Melting Point 48°C to 50°C
Flash Point >110°C (230°F)
Quantity 5 g
Beilstein 1239364
Formula Weight 166.18
Percent Purity 99%
Chemical Name or Material 2'-Hydroxy-5'-methoxyacetophenone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 211-882-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only