missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-5-iodopyridine 98.0+%, TCI America™

Catalog No. H14661G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H14661G 1 g
H14665G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H14661G Supplier TCI America Supplier No. H14661G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 13472-79-2 | C5H4INO | 220.997 g/mol

Chemical Identifiers

CAS 13472-79-2
Molecular Formula C5H4INO
Molecular Weight (g/mol) 220.997
MDL Number MFCD00234058
InChI Key ZDJUNNCVIDKJAN-UHFFFAOYSA-N
Synonym 2-hydroxy-5-iodopyridine, 5-iodopyridin-2-ol, 5-iodopyridin-2 1h-one, 5-iodo-pyridin-2-ol, 5-iodo-2-pyridone, 2 1h-pyridinone, 5-iodo, 5-iodo-2-pyridinol, 5-jod-2-pyridon, 5-iodo-2-pyridol, 5-iodopyridin-2-one
PubChem CID 459500
IUPAC Name 5-iodo-1H-pyridin-2-one
SMILES C1=CC(=O)NC=C1I

Specifications

Melting Point 190°C
Color White-Yellow
Quantity 1 g
Formula Weight 221.00
Percent Purity ≥98.0% (GC,T)
Grade Not Available
Physical Form Crystalline Powder
Chemical Name or Material 2-Hydroxy-5-iodopyridine