missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-5-iodopyridine, 97%

Catalog No. p-7047979
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL2004403 1 g
AAL2004406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL2004403 Supplier Thermo Scientific Chemicals Supplier No. L2004403

CAS: 13472-79-2 | C5H4INO | 220.997 g/mol

Employed as a important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Employed as a important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Light Sensitive. Store in dark. Store in cool, dry place in tightly closed container. Store away from oxidizing agent.

Chemical Identifiers

CAS 13472-79-2
Molecular Formula C5H4INO
Molecular Weight (g/mol) 220.997
MDL Number MFCD00234058
InChI Key ZDJUNNCVIDKJAN-UHFFFAOYSA-N
Synonym 2-hydroxy-5-iodopyridine, 5-iodopyridin-2-ol, 5-iodopyridin-2 1h-one, 5-iodo-pyridin-2-ol, 5-iodo-2-pyridone, 2 1h-pyridinone, 5-iodo, 5-iodo-2-pyridinol, 5-jod-2-pyridon, 5-iodo-2-pyridol, 5-iodopyridin-2-one
PubChem CID 459500
IUPAC Name 5-iodo-1H-pyridin-2-one
SMILES C1=CC(=O)NC=C1I

Specifications

Melting Point 189°C to 191°C
Quantity 1 g
Beilstein 108752
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 221
Percent Purity 97%
Chemical Name or Material 2-Hydroxy-5-iodopyridine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only