missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-5-bromopyridine, 99%, Thermo Scientific™

Chemical Identifiers

CAS 13466-38-1
Molecular Formula C5H4BrNO
Molecular Weight (g/mol) 174
MDL Number MFCD00234042
InChI Key NDMZZQRNZFWMEZ-UHFFFAOYSA-N
Synonym 5-bromo-2-hydroxypyridine, 5-bromopyridin-2-ol, 2-hydroxy-5-bromopyridine, 5-bromopyridin-2 1h-one, 5-bromo-2-pyridinol, 5-bromo-2 1h-pyridinone, 5-bromo-2 1h-pyridone, 2 1h-pyridinone, 5-bromo, 3-bromo-6-hydroxypyridine, 5-bromo-1,2-dihydropyridin-2-one
PubChem CID 599528
IUPAC Name 5-bromo-1H-pyridin-2-one
SMILES C1=CC(=O)NC=C1Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
AC357640050
View Documents Promotion Details
N/A 5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC357640250
View Documents Promotion Details
N/A 25g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

13466-38-1
174
NDMZZQRNZFWMEZ-UHFFFAOYSA-N
599528
C1=CC(=O)NC=C1Br
C5H4BrNO
MFCD00234042
5-bromo-2-hydroxypyridine, 5-bromopyridin-2-ol, 2-hydroxy-5-bromopyridine, 5-bromopyridin-2 1h-one, 5-bromo-2-pyridinol, 5-bromo-2 1h-pyridinone, 5-bromo-2 1h-pyridone, 2 1h-pyridinone, 5-bromo, 3-bromo-6-hydroxypyridine, 5-bromo-1,2-dihydropyridin-2-one
5-bromo-1H-pyridin-2-one

Specifications

13466-38-1
100.0
98.5% min. (GC)
MFCD00234042
NDMZZQRNZFWMEZ-UHFFFAOYSA-N
5-bromo-1H-pyridin-2-one
599528
99%
98.5
Authentic
C5H4BrNO
5-bromo-2-hydroxypyridine, 5-bromopyridin-2-ol, 2-hydroxy-5-bromopyridine, 5-bromopyridin-2 1h-one, 5-bromo-2-pyridinol, 5-bromo-2 1h-pyridinone, 5-bromo-2 1h-pyridone, 2 1h-pyridinone, 5-bromo, 3-bromo-6-hydroxypyridine, 5-bromo-1,2-dihydropyridin-2-one
C1=CC(=O)NC=C1Br
174
174
2-Hydroxy-5-bromopyridine, 99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents

RUO – Research Use Only