missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-4-(trifluoromethyl)pyrimidine 97.0+%, TCI America™

Catalog No. p-7130982
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H13281G 1 g
H13285G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H13281G Supplier TCI America Supplier No. H13281G
Only null left

Chemical Identifiers

CAS 104048-92-2
Molecular Formula C5H3F3N2O
Molecular Weight (g/mol) 164.09
MDL Number MFCD00192529
InChI Key WCEBFEVZTGLOHC-UHFFFAOYSA-N
Synonym 2-hydroxy-4-trifluoromethyl pyrimidine, 4-trifluoromethyl pyrimidin-2-ol, 4-trifluoromethyl pyrimidin-2 1h-one, 2-hydroxy-4-trifluoromethylpyrimidine, 4-trifluoromethyl-2-pyrimidinol, 6-trifluoromethyl-1h-pyrimidin-2-one, 2 1h-pyrimidinone, 4-trifluoromethyl, pubchem7201, enamine_002404, acmc-2098am
PubChem CID 1240050
IUPAC Name 6-(trifluoromethyl)-1,2-dihydropyrimidin-2-one
SMILES FC(F)(F)C1=CC=NC(=O)N1

Specifications

Melting Point 219°C
Color White-Yellow
Quantity 1 g
Formula Weight 164.09
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Hydroxy-4-(trifluoromethyl)pyrimidine