missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-4-methylvaleric acid, 98%

Catalog No. AAH6045903
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6045903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6045903 Supplier Thermo Scientific Chemicals Supplier No. H6045903
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 498-36-2 | C6H12O3 | 132.159 g/mol

2-Hydroxy-4-methylvaleric acid is used as nutrients, stabilizers, surfactants and emulsifiers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Hydroxy-4-methylvaleric acid is used as nutrients, stabilizers, surfactants and emulsifiers.

Solubility
Soluble in water. (82 g/L) at 25°C.

Notes
Store in a cool, dry conditions in a well seal

Chemical Identifiers

CAS 498-36-2
Molecular Formula C6H12O3
Molecular Weight (g/mol) 132.159
MDL Number MFCD00004246
InChI Key LVRFTAZAXQPQHI-UHFFFAOYSA-N
Synonym 2-hydroxy-4-methylvaleric acid, 2-hydroxyisocaproic acid, leucic acid, dl-leucic acid, alpha-hydroxyisocaproic acid, leucinic acid, 2-hydroxy-4-methyl-pentanoic acid, pentanoic acid, 2-hydroxy-4-methyl, dl-2-hydroxyisocaproic acid, leucate
PubChem CID 92779
ChEBI CHEBI:59783
IUPAC Name 2-hydroxy-4-methylpentanoic acid
SMILES CC(C)CC(C(=O)O)O

Specifications

Melting Point 78°C to 80°C
Linear Formula (CH3)2CHCH2CH(OH)CO2H
Quantity 1 g
Solubility Information Soluble in water. (82g/L) at 25°C.
Formula Weight 132.16
Percent Purity 98%
Chemical Name or Material 2-Hydroxy-4-methylvaleric acid
EINECSNumber 207-860-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only