missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-4-methylpyridine 98.0+%, TCI America™

Catalog No. H10525G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H10525G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H10525G Supplier TCI America Supplier No. H10525G

Chemical Identifiers

CAS 13466-41-6
Molecular Formula C6H7NO
Molecular Weight (g/mol) 109.13
MDL Number MFCD00134298,MFCD00030766
InChI Key YBDRFJXGJQULGH-UHFFFAOYSA-N
Synonym 2-hydroxy-4-methylpyridine, 4-methylpyridin-2-ol, 4-methyl-2 1h-pyridinone, 2 1h-pyridinone, 4-methyl, 2-hydroxy-4-picoline, 4-methyl-2-pyridinol, 4-methyl-1,2-dihydropyridin-2-one, 4-methyl-2-pyridone, 4-methylpyridin-2 1h-one, 2-hydroxy-4-methyl pyridine
PubChem CID 280741
IUPAC Name 4-methyl-1,2-dihydropyridin-2-one
SMILES CC1=CC(=O)NC=C1

Specifications

Melting Point 130°C
Boiling Point 309°C
Quantity 5 g
Formula Weight 109.13
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Hydroxy-4-methylpyridine