missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-4-methylpyridine, 98+%

Catalog No. p-7046749
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1618803 1 g
AAL1618806 5 g
AAL1618814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1618803 Supplier Thermo Scientific Chemicals Supplier No. L1618803
Only null left

CAS: 13466-41-6 | C6H7NO | 109.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13466-41-6
Molecular Formula C6H7NO
Molecular Weight (g/mol) 109.13
MDL Number MFCD00134298,MFCD00030766
InChI Key YBDRFJXGJQULGH-UHFFFAOYSA-N
Synonym 2-hydroxy-4-methylpyridine, 4-methylpyridin-2-ol, 4-methyl-2 1h-pyridinone, 2 1h-pyridinone, 4-methyl, 2-hydroxy-4-picoline, 4-methyl-2-pyridinol, 4-methyl-1,2-dihydropyridin-2-one, 4-methyl-2-pyridone, 4-methylpyridin-2 1h-one, 2-hydroxy-4-methyl pyridine
PubChem CID 280741
SMILES CC1=CC(=O)NC=C1

Specifications

Melting Point 128°C to 131°C
Boiling Point 186°C to 187°C (12 mmHg)
Odor Odorless
Quantity 1 g
Beilstein 107082
IUPAC Name 4-methyl-1H-pyridin-2-one
Formula Weight 109.13
Percent Purity ≥98%
Chemical Name or Material 2-Hydroxy-4-methylpyridine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only