missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-4-methylbenzaldehyde, 97+%

Catalog No. AAH3390914
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3390914 25 g
AAH3390906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3390914 Supplier Thermo Scientific Chemicals Supplier No. H3390914
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 698-27-1 | C8H8O2 | 136.15 g/mol

The intermediate for synthesis of liquid crystal. Also used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The intermediate for synthesis of liquid crystal. Also used as pharmaceutical intermediate.

Solubility
Soluble in oil, slightly soluble in water.

Notes
Air sensitive. Store away from air, oxidizing agents. Store under inert gas.

Chemical Identifiers

CAS 698-27-1
Molecular Formula C8H8O2
Molecular Weight (g/mol) 136.15
MDL Number MFCD00799550
InChI Key JODRRPJMQDFCBJ-UHFFFAOYSA-N
Synonym 4-methylsalicylaldehyde, 2,4-cresotaldehyde, benzaldehyde, 2-hydroxy-4-methyl, 4-methyl-2-hydroxybenzaldehyde, 2-hydroxy-4-methyl-benzaldehyde, 2-formyl-5-methylphenol, 4-methylsalicyclic aldehyde, unii-9n0s26g61j, 4-methyl salicylaldehyde, homosalicylaldehyde, m
PubChem CID 61200
IUPAC Name 2-hydroxy-4-methylbenzaldehyde
SMILES CC1=CC=C(C=O)C(O)=C1

Specifications

Melting Point 58°C to 62°C
Boiling Point 222°C to 223°C
Quantity 25 g
Sensitivity Air Sensitive
Solubility Information Soluble in oil,slightly soluble in water.
Formula Weight 136.15
Percent Purity ≥97%
Chemical Name or Material 2-Hydroxy-4-methylbenzaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 211-813-7
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only