missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-4-methyl-5-nitropyridine, 98%, Thermo Scientific™

Catalog No. p-3758202
Change view
Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC319570010 1g Glass bottle
AC319570050 5g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC319570010 Supplier Thermo Scientific Chemicals Supplier No. 319570010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 21901-41-7
Molecular Formula C6H6N2O3
Molecular Weight (g/mol) 154.13
MDL Number MFCD00010690
InChI Key AIEHUZHKFUNHCJ-UHFFFAOYSA-N
Synonym 2-hydroxy-4-methyl-5-nitropyridine, 4-methyl-5-nitropyridin-2-ol, 2-hydroxy-5-nitro-4-picoline, 2-hydroxy-5-nitro-4-methyl pyridine, 2-hydroxy-5-nitro-4-methylpyridine, 4-methyl-5-nitropyridin-2 1h-one, 4-methyl-5-nitro-2-pyridinol, 4-methyl-5-nitro-1,2-dihydropyridin-2-one, 4-methyl-5-nitro-pyridin-2-ol, 2 1h-pyridinone, 4-methyl-5-nitro
PubChem CID 345371
IUPAC Name 4-methyl-5-nitro-1H-pyridin-2-one
SMILES CC1=CC(=O)NC=C1[N+](=O)[O-]

Specifications

CAS Min % 97.5
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 1g
Formula Weight 154.13
Percent Purity 98%
Chemical Name or Material 2-Hydroxy-4-methyl-5-nitropyridine
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Harmful if inhaled.
Harmful in contact with skin.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only