missing translation for 'onlineSavingsMsg'
Learn More

2'-Hydroxy-4'-methoxyacetophenone 98.0+%, TCI America™

Catalog No. H07895G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H07895G 5 g
H078925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H07895G Supplier TCI America Supplier No. H07895G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 552-41-0
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00008730
InChI Key UILPJVPSNHJFIK-UHFFFAOYSA-N
Synonym paeonol, 2'-hydroxy-4'-methoxyacetophenone, 1-2-hydroxy-4-methoxyphenyl ethanone, peonol, 2-hydroxy-4-methoxyacetophenone, 1-2-hydroxy-4-methoxyphenyl ethan-1-one, resacetophenone-4-methyl ether, ethanone, 1-2-hydroxy-4-methoxyphenyl, paeonal, paeonolum
PubChem CID 11092
ChEBI CHEBI:69581
IUPAC Name 1-(2-hydroxy-4-methoxyphenyl)ethanone
SMILES CC(=O)C1=C(C=C(C=C1)OC)O

Specifications

Melting Point 50°C
Color White
Quantity 5 g
Formula Weight 166.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2′-Hydroxy-4′-methoxyacetophenone
EINECSNumber (3)-1252
RTECSNumber RT1215000
TSCA No