missing translation for 'onlineSavingsMsg'
Learn More

2'-Hydroxy-4'-methoxyacetophenone, 99%

Catalog No. p-7026259
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2217306 5 g
AAB2217314 25 g
AAB2217322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2217306 Supplier Thermo Scientific Chemicals Supplier No. B2217306

CAS: 552-41-0 | C9H10O3 | 166.176 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 552-41-0
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00008730
InChI Key UILPJVPSNHJFIK-UHFFFAOYSA-N
Synonym paeonol, 2'-hydroxy-4'-methoxyacetophenone, 1-2-hydroxy-4-methoxyphenyl ethanone, peonol, 2-hydroxy-4-methoxyacetophenone, 1-2-hydroxy-4-methoxyphenyl ethan-1-one, resacetophenone-4-methyl ether, ethanone, 1-2-hydroxy-4-methoxyphenyl, paeonal, paeonolum
PubChem CID 11092
ChEBI CHEBI:69581
IUPAC Name 1-(2-hydroxy-4-methoxyphenyl)ethanone
SMILES CC(=O)C1=C(C=C(C=C1)OC)O

Specifications

Melting Point 48°C to 50°C
Flash Point >110°C (230°F)
Quantity 5 g
Beilstein 1282794
Formula Weight 166.18
Percent Purity 99%
Chemical Name or Material 2'-Hydroxy-4'-methoxyacetophenone
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 209-012-2
RTECSNumber RT1215000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only