Learn More
2'-Hydroxy-4'-methoxyacetophenone, 99%
CAS: 552-41-0 | C9H10O3 | 166.176 g/mol
$155.24 - $155.24
Chemical Identifiers
| CAS | 552-41-0 |
|---|---|
| MDL Number | MFCD00008730 |
| InChI Key | UILPJVPSNHJFIK-UHFFFAOYSA-N |
| Synonym | paeonol, 2'-hydroxy-4'-methoxyacetophenone, 1-2-hydroxy-4-methoxyphenyl ethanone, peonol, 2-hydroxy-4-methoxyacetophenone, 1-2-hydroxy-4-methoxyphenyl ethan-1-one, resacetophenone-4-methyl ether, ethanone, 1-2-hydroxy-4-methoxyphenyl, paeonal, paeonolum |
| PubChem CID | 11092 |
| ChEBI | CHEBI:69581 |
| IUPAC Name | 1-(2-hydroxy-4-methoxyphenyl)ethanone |
| SMILES | CC(=O)C1=C(C=C(C=C1)OC)O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC121330100
|
Thermo Scientific Chemicals
121330100 |
10 g | Glass bottle |
Each for $155.24
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 552-41-0 | |
| UILPJVPSNHJFIK-UHFFFAOYSA-N | |
| 11092 | |
| 1-(2-hydroxy-4-methoxyphenyl)ethanone |
| MFCD00008730 | |
| paeonol, 2'-hydroxy-4'-methoxyacetophenone, 1-2-hydroxy-4-methoxyphenyl ethanone, peonol, 2-hydroxy-4-methoxyacetophenone, 1-2-hydroxy-4-methoxyphenyl ethan-1-one, resacetophenone-4-methyl ether, ethanone, 1-2-hydroxy-4-methoxyphenyl, paeonal, paeonolum | |
| CHEBI:69581 | |
| CC(=O)C1=C(C=C(C=C1)OC)O |
Specifications
| 552-41-0 | |
| 100.0 | |
| Beige to White | |
| 99% | |
| C9H10O3 | |
| 10 g | |
| paeonol, 2'-hydroxy-4'-methoxyacetophenone, 1-2-hydroxy-4-methoxyphenyl ethanone, peonol, 2-hydroxy-4-methoxyacetophenone, 1-2-hydroxy-4-methoxyphenyl ethan-1-one, resacetophenone-4-methyl ether, ethanone, 1-2-hydroxy-4-methoxyphenyl, paeonal, paeonolum | |
| CC(=O)C1=C(C=C(C=C1)OC)O | |
| 166.176 | |
| CHEBI:69581 | |
| ≥98.5% | |
| 2'-Hydroxy-4'-methoxyacetophenone |
| 98.5 | |
| 47°C to 50°C | |
| Authentic | |
| Glass bottle | |
| MFCD00008730 | |
| 08, 267 | |
| UILPJVPSNHJFIK-UHFFFAOYSA-N | |
| 1-(2-hydroxy-4-methoxyphenyl)ethanone | |
| 11092 | |
| 166.18 | |
| Crystalline Powder |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
RUO – Research Use Only