missing translation for 'onlineSavingsMsg'
Learn More

2'-Hydroxy-4',5'-dimethylacetophenone 98.0+%, TCI America™

Catalog No. D48125G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D48125G 5 g
D48121G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D48125G Supplier TCI America Supplier No. D48125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 36436-65-4
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.204
MDL Number MFCD00002306
InChI Key YXVSURZEXVMUAM-UHFFFAOYSA-N
PubChem CID 118976
IUPAC Name 1-(2-hydroxy-4,5-dimethylphenyl)ethanone
SMILES CC1=C(C=C(C(=C1)C(=O)C)O)C

Specifications

Melting Point 72°C
Color White-Yellow
Boiling Point 144°C
Quantity 5 g
Synonym 2'-hydroxy-4',5'-dimethylacetophenone, 1-2-hydroxy-4,5-dimethylphenyl ethanone, 2-hydroxy-4,5-dimethylacetophenone, 4'5'-dimethyl-2'-hydroxyacetophenone, ethanone, 1-2-hydroxy-4,5-dimethylphenyl, 4',5'-dimethyl-2'-hydroxyacetophenone, 1-2-hydroxy-4,5-dimethylphenyl ethan-1-one, 1-acetyl-2-hydroxy-4,5-dimethylbenzene, ethanone,1-2-hydroxy-4,5-dimethylphenyl, acmc-1ctvf
Formula Weight 164.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2′-Hydroxy-4′,5′-dimethylacetophenone