Learn More
2'-Hydroxy-4',5'-dimethylacetophenone, 98%
CAS: 36436-65-4 | C10H12O2 | 164.204 g/mol
$45.90 - $154.30
Chemical Identifiers
| CAS | 36436-65-4 |
|---|---|
| Molecular Formula | C10H12O2 |
| Molecular Weight (g/mol) | 164.204 |
| MDL Number | MFCD00002306 |
| InChI Key | YXVSURZEXVMUAM-UHFFFAOYSA-N |
| PubChem CID | 118976 |
| IUPAC Name | 1-(2-hydroxy-4,5-dimethylphenyl)ethanone |
| SMILES | CC1=C(C=C(C(=C1)C(=O)C)O)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH5068503
|
Thermo Scientific Chemicals
H5068503 |
1 g |
Each for $45.90
|
|
|||||
|
AAH5068506
|
Thermo Scientific Chemicals
H5068506 |
5 g |
Each for $154.30
|
|
|||||
Description
2'-Hydroxy-4',5'-dimethylacetophenone is used in the preparation of chalcone derivatives with antibacterial activity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2′-Hydroxy-4′,5′-dimethylacetophenone is used in the preparation of chalcone derivatives with antibacterial activity.
Solubility
Slightly soluble in water.
Notes
Store in a cool, dry condition in a well sealed container. Incompatible with oxidizing agents.
Chemical Identifiers
| 36436-65-4 | |
| 164.204 | |
| YXVSURZEXVMUAM-UHFFFAOYSA-N | |
| 1-(2-hydroxy-4,5-dimethylphenyl)ethanone |
| C10H12O2 | |
| MFCD00002306 | |
| 118976 | |
| CC1=C(C=C(C(=C1)C(=O)C)O)C |
Specifications
| 36436-65-4 | |
| C10H12O2 | |
| 1 g | |
| Slightly soluble in water. | |
| CC1=C(C=C(C(=C1)C(=O)C)O)C | |
| 164.204 | |
| 164.21 | |
| 2'-Hydroxy-4',5'-dimethylacetophenone |
| 88°C to 90°C | |
| MFCD00002306 | |
| 2'-hydroxy-4',5'-dimethylacetophenone, 1-2-hydroxy-4,5-dimethylphenyl ethanone, 2-hydroxy-4,5-dimethylacetophenone, 4'5'-dimethyl-2'-hydroxyacetophenone, ethanone, 1-2-hydroxy-4,5-dimethylphenyl, 4',5'-dimethyl-2'-hydroxyacetophenone, 1-2-hydroxy-4,5-dimethylphenyl ethan-1-one, 1-acetyl-2-hydroxy-4,5-dimethylbenzene, ethanone,1-2-hydroxy-4,5-dimethylphenyl, acmc-1ctvf | |
| YXVSURZEXVMUAM-UHFFFAOYSA-N | |
| 1-(2-hydroxy-4,5-dimethylphenyl)ethanone | |
| 118976 | |
| 98% |
Safety and Handling
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 253-035-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only