missing translation for 'onlineSavingsMsg'
Learn More

2'-Hydroxy-4',5'-dimethylacetophenone, 98%

Catalog No. AAH5068506
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5068506 5 g
AAH5068503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5068506 Supplier Thermo Scientific Chemicals Supplier No. H5068506

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 36436-65-4 | C10H12O2 | 164.204 g/mol

2'-Hydroxy-4',5'-dimethylacetophenone is used in the preparation of chalcone derivatives with antibacterial activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′-Hydroxy-4′,5′-dimethylacetophenone is used in the preparation of chalcone derivatives with antibacterial activity.

Solubility
Slightly soluble in water.

Notes
Store in a cool, dry condition in a well sealed container. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 36436-65-4
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.204
MDL Number MFCD00002306
InChI Key YXVSURZEXVMUAM-UHFFFAOYSA-N
PubChem CID 118976
IUPAC Name 1-(2-hydroxy-4,5-dimethylphenyl)ethanone
SMILES CC1=C(C=C(C(=C1)C(=O)C)O)C

Specifications

Melting Point 88°C to 90°C
Quantity 5 g
Synonym 2'-hydroxy-4',5'-dimethylacetophenone, 1-2-hydroxy-4,5-dimethylphenyl ethanone, 2-hydroxy-4,5-dimethylacetophenone, 4'5'-dimethyl-2'-hydroxyacetophenone, ethanone, 1-2-hydroxy-4,5-dimethylphenyl, 4',5'-dimethyl-2'-hydroxyacetophenone, 1-2-hydroxy-4,5-dimethylphenyl ethan-1-one, 1-acetyl-2-hydroxy-4,5-dimethylbenzene, ethanone,1-2-hydroxy-4,5-dimethylphenyl, acmc-1ctvf
Solubility Information Slightly soluble in water.
Formula Weight 164.21
Percent Purity 98%
Chemical Name or Material 2'-Hydroxy-4',5'-dimethylacetophenone
Hazard Category H302-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 253-035-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only