missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Hydroxy-4'-(2-hydroxyethoxy)-2-methylpropiophenone 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 91°C |
| Color | White-Yellow |
| Quantity | 5 g |
| Formula Weight | 224.26 |
| Percent Purity | ≥98.0% (HPLC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2-Hydroxy-4′-(2-hydroxyethoxy)-2-methylpropiophenone |
Safety and Handling
| EINECSNumber | (3)-4272 |
| RTECSNumber | UC2976000 |
| TSCA | Yes |