missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-3-methylbenzaldehyde, 98%

Catalog No. AC398540010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC398540010 1 g Glass bottle
AC398540050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC398540010 Supplier Thermo Scientific Chemicals Supplier No. 398540010

CAS: 824-42-0 | C8H8O2 | 136.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 824-42-0
Molecular Formula C8H8O2
Molecular Weight (g/mol) 136.15
MDL Number MFCD00040798
InChI Key IPPQNXSAJZOTJZ-UHFFFAOYSA-N
Synonym 3-methylsalicylaldehyde, 2,3-cresotaldehyde, 2-hydroxy-3-methyl-benzaldehyde, 3-methylsalicylic aldehyde, 3-methyl-2-hydroxybenzaldehyde, benzaldehyde, hydroxy-3-methyl, benzaldehyde, 2-hydroxy-3-methyl, zlchem 747, pubchem14264, 2-formyl-6-methylphenol
PubChem CID 522777
ChEBI CHEBI:20110
IUPAC Name 2-hydroxy-3-methylbenzaldehyde
SMILES CC1=CC=CC(=C1O)C=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Orange to Yellow
Density 1.1340g/mL
Boiling Point 203°C to 205°C (730.0 mmHg)
Flash Point 87°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3C6H3(OH)CHO
Quantity 1 g
Specific Gravity 1.134
Formula Weight 136.15
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Hydroxy-3-methylbenzaldehyde
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only