missing translation for 'onlineSavingsMsg'
Learn More

2'-Hydroxy-3',4'-dimethoxyacetophenone 98.0+%, TCI America™

Catalog No. p-7130922
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H12055G 5 g
H120525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H12055G Supplier TCI America Supplier No. H12055G

Chemical Identifiers

CAS 5396-18-9
Molecular Formula C10H12O4
Molecular Weight (g/mol) 196.20
MDL Number MFCD00016684
InChI Key BCEPNLMYVYJIHU-UHFFFAOYSA-N
PubChem CID 219639
IUPAC Name 1-(2-hydroxy-3,4-dimethoxyphenyl)ethan-1-one
SMILES COC1=CC=C(C(C)=O)C(O)=C1OC

Specifications

Melting Point 77°C
Quantity 5 g
Formula Weight 196.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2′-Hydroxy-3′,4′-dimethoxyacetophenone