missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-2-(trifluoromethyl)propionitrile 96.0+%, TCI America™

Catalog No. H15081G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H15081G 1 g
H15085G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H15081G Supplier TCI America Supplier No. H15081G

Chemical Identifiers

CAS 335-08-0
Molecular Formula C4H4F3NO
Molecular Weight (g/mol) 139.077
MDL Number MFCD00040885
InChI Key XDCMNDCKYSQKAX-UHFFFAOYSA-N
Synonym 1,1,1-trifluoroacetone cyanohydrin, 2-cyano-1,1,1-trifluoro-2-propanol, 2-hydroxy-2-trifluoromethyl propionitrile, propanenitrile, 3,3,3-trifluoro-2-hydroxy-2-methyl, 2-trifluoromethyl lactonitrile, trifluoroacetone cyanohydrin, lactonitrile, 2-trifluoromethyl, 3-trifluoro-a-hydroxyisobutyronitrile, lactonitrile,3,3-trifluoro-2-methyl, 2-cyano-1,1,1-trifluoropropan-2-ol
PubChem CID 274400
IUPAC Name 3,3,3-trifluoro-2-hydroxy-2-methylpropanenitrile
SMILES CC(C#N)(C(F)(F)F)O

Specifications

Color Colorless
Boiling Point 139°C
Quantity 1 g
UN Number 3275
Formula Weight 139.08
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Hydroxy-2-(trifluoromethyl)propionitrile