Learn More
2-Hydroxy-2-(trifluoromethyl)propionic acid, 94%
CAS: 374-35-6 | C4H5F3O3 | 158.08 g/mol
$220.66 - $220.66
Chemical Identifiers
| CAS | 374-35-6 |
|---|---|
| Molecular Formula | C4H5F3O3 |
| Molecular Weight (g/mol) | 158.08 |
| MDL Number | MFCD00190646,MFCD03095400,MFCD07375449 |
| InChI Key | CTGJACFEVDCYMC-UHFFFAOYNA-N |
| Synonym | 2-trifluoromethyl-2-hydroxypropionic acid, 3,3,3-trifluoro-2-hydroxy-2-methylpropionic acid, 2-hydroxy-2-trifluoromethyl propionic acid, 3,3,3-trifluoro-2-hydroxy-2-methyl-propionic acid, propanoic acid, 3,3,3-trifluoro-2-hydroxy-2-methyl, methyl3,3,3-trifluoro-dl-lactate, acmc-1ai3k, acmc-209gc9, 2s-2-hydroxy-2-methyl-3,3,3-trifluoropropanoic acid, alpha-trifluoromethyl lactic acid |
| PubChem CID | 2775125 |
| IUPAC Name | 3,3,3-trifluoro-2-hydroxy-2-methylpropanoic acid |
| SMILES | CC(O)(C(O)=O)C(F)(F)F |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL1739906
|
Thermo Scientific Chemicals
L1739906 |
5 g |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 374-35-6 | |
| 158.08 | |
| CTGJACFEVDCYMC-UHFFFAOYNA-N | |
| 2775125 | |
| CC(O)(C(O)=O)C(F)(F)F |
| C4H5F3O3 | |
| MFCD00190646,MFCD03095400,MFCD07375449 | |
| 2-trifluoromethyl-2-hydroxypropionic acid, 3,3,3-trifluoro-2-hydroxy-2-methylpropionic acid, 2-hydroxy-2-trifluoromethyl propionic acid, 3,3,3-trifluoro-2-hydroxy-2-methyl-propionic acid, propanoic acid, 3,3,3-trifluoro-2-hydroxy-2-methyl, methyl3,3,3-trifluoro-dl-lactate, acmc-1ai3k, acmc-209gc9, 2s-2-hydroxy-2-methyl-3,3,3-trifluoropropanoic acid, alpha-trifluoromethyl lactic acid | |
| 3,3,3-trifluoro-2-hydroxy-2-methylpropanoic acid |
Specifications
| 374-35-6 | |
| 189°C | |
| MFCD00190646,MFCD03095400,MFCD07375449 | |
| UN3261 | |
| 2-trifluoromethyl-2-hydroxypropionic acid, 3,3,3-trifluoro-2-hydroxy-2-methylpropionic acid, 2-hydroxy-2-trifluoromethyl propionic acid, 3,3,3-trifluoro-2-hydroxy-2-methyl-propionic acid, propanoic acid, 3,3,3-trifluoro-2-hydroxy-2-methyl, methyl3,3,3-trifluoro-dl-lactate, acmc-1ai3k, acmc-209gc9, 2s-2-hydroxy-2-methyl-3,3,3-trifluoropropanoic acid, alpha-trifluoromethyl lactic acid | |
| CC(O)(C(O)=O)C(F)(F)F | |
| 158.08 | |
| 158.08 | |
| 2-Hydroxy-2-(trifluoromethyl)propionic acid |
| ∼85°C | |
| C4H5F3O3 | |
| 5 g | |
| 1723691 | |
| CTGJACFEVDCYMC-UHFFFAOYNA-N | |
| 3,3,3-trifluoro-2-hydroxy-2-methylpropanoic acid | |
| 2775125 | |
| 94% |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Hazard Class: 8; Packaging Group: III
EINECSNumber : 609-428-8
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only